missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ RTF1 Antibody [FITC], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NB600-277F
This item is not returnable.
View return policy
Description
RTF1 Polyclonal antibody specifically detects RTF1 in Human samples. It is validated for Western Blot,Immunoprecipitation (Negative)
Specifications
| RTF1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation (Negative) | |
| FITC | |
| KIAA0252GTL7, ortholog of mouse gene trap locus 7, RNA polymerase-associated protein RTF1 homolog, Rtf1, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae) | |
| The immunogen recognized by this antibody maps to a region between residues 300 and 350 of human Restores TBP (TATA box-binding protein) function-1 using the numbering given in entry EAW92492.1 (GeneID 23168). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 23168 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction