missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribonuclease A Antibody (058), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90219CL1
This item is not returnable.
View return policy
Description
Ribonuclease A Monoclonal antibody specifically detects Ribonuclease A in Human samples. It is validated for ELISA
Specifications
| Ribonuclease A | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 3.1.27.5, HP-RNase, MGC12408, RIB1, RIB-1, Ribonuclease 1, Ribonuclease A, ribonuclease pancreatic, ribonuclease, RNase A family, 1 (pancreatic), RNase A, RNase UpI-1, RNS1RNase 1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Ribonuclease A (Uniprot#: P07998; Met1-Thr156). | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing, Proteases & Other Enzymes | |
| 6035 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| 058 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction