missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGL2 Antibody (OTI10F2), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73875CL1
This item is not returnable.
View return policy
Description
RGL2 Monoclonal antibody specifically detects RGL2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| RGL2 | |
| Monoclonal | |
| CoraFluor 1 | |
| GDS-related protein, HKE1.5, KE1.5, RAB2, member RAS oncogene family-like, RAB2LRas-associated protein RAB2L, ral guanine nucleotide dissociation stimulator-like 2, RalGDS-like 2, RalGDS-like factor | |
| Human recombinant protein fragment corresponding to amino acids 547-777 of human RGL2(NP_004752) produced in E.coli. | |
| 0.1 mL | |
| Signal Transduction | |
| 5863 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Immunohistochemistry, Western Blot | |
| OTI10F2 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction