missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ VGLL3 Antibody [Biotin], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-26910B
This item is not returnable.
View return policy
Description
VGLL3 Polyclonal antibody specifically detects VGLL3 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| VGLL3 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 389136 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Biotin | |
| colon carcinoma related protein, DKFZp686O1845, FLJ38507, transcription cofactor vestigial-like protein 3, vestigial like 3 (Drosophila), vestigial-like 3, Vgl-3, VGL3 | |
| Produced in rabbits immunized with E. coli-derived Human VGLL3 fragment | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction