missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LEDGF Antibody [PerCP], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NB110-40560PCP
This item is not returnable.
View return policy
Description
LEDGF Polyclonal antibody specifically detects LEDGF in Human,Mouse samples. It is validated for Immunohistochemistry,Immunoprecipitation,Immunohistochemistry (Paraffin),Western Blot (Negative)
Specifications
| LEDGF | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot (Negative) | |
| PerCP | |
| CLL-associated antigen KW-7, Dense fine speckles 70 kDa protein, DFS70, LEDGFDFS 70, Lens epithelium-derived growth factor, MGC74712, p52, p75, PAIP, PC4 and SFRS1 interacting protein 1, PC4 and SFRS1 interacting protein 2, PC4 and SFRS1-interacting protein, PSIP2, transcriptional coactivator p52/p75, Transcriptional coactivator p75/p52 | |
| The immunogen recognized by this antibody maps to a region between residue 225 and 275 of human Lens Epithelium Derived Growth Factor/Transcriptional Coactivator p75 using the numbering given in entry NP_150091.2 (GeneID 11168). | |
| 0.1 mL | |
| ABC Transporters, Alzheimers Research, Neuroscience | |
| 11168 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction