missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIGNL1 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-26083PE
This item is not returnable.
View return policy
Description
FIGNL1 Polyclonal antibody specifically detects FIGNL1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| FIGNL1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 3.6.4, EC 3.6.4.-, fidgetin-like 1, fidgetin-like protein 1 | |
| Produced in rabbits immunized with E. coli-derived Human FIGNL1 fragment | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 63979 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction