missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FAM184B Antibody [PE], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NB300-242PE
This item is not returnable.
View return policy
Description
FAM184B Polyclonal antibody specifically detects FAM184B in Human samples. It is validated for Western Blot
Specifications
| FAM184B | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| PE | |
| family with sequence similarity 184, member B, hypothetical protein LOC27146, KIAA1276 | |
| A synthetic peptide mapping to a region between residue 1000 and the C-terminus (residue 1068) of human KIAA1276 using the numbering given in TrEMBL entry Q9ULE4 (GeneID 27146). | |
| 0.1 mL | |
| Neuroscience | |
| 27146 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction