missing translation for 'onlineSavingsMsg'
Learn More

PTP pi/PTPRU Antibody (PTPRU/7616), HRP, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-24115H

Product Code. 18605004

  • £514.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PTP pi/PTPRU Monoclonal antibody specifically detects PTP pi/PTPRU in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PTP pi/PTPRU
Monoclonal
HRP
FMI, hPTP-J, pancreatic carcinoma phosphatase 2, PCP2, PCP-2, pi R-PTP-Psi, protein tyrosine phosphatase J, protein tyrosine phosphatase, receptor type, U, protein-tyrosine phosphatase J, protein-tyrosine phosphatase pi, PTP, PTP pi, PTP-J, PTP-PI, PTPPSI, PTP-RO, Receptor protein tyrosine phosphatase hPTP-J, R-PTP-PSI
Recombinant fragment (around aa1-200) of human PTP pi/PTPRU protein (exact sequence is proprietary)
0.1 mL
Signal Transduction
10076
Store at 4°C in the dark.
IgG2 κ
Immunohistochemistry (Paraffin)
PTPRU/7616
PBS
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.