Learn More
PGLYRP2/PGRP-L Antibody (439728), Alexa Fluor™ 405, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB4704V
Product Code. 30062272
  • £452.00 / 0.10mL
Description
PGLYRP2/PGRP-L Monoclonal antibody specifically detects PGLYRP2/PGRP-L in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PGLYRP2/PGRP-L
Monoclonal
Alexa Fluor 405
50mM Sodium Borate
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Mouse
Purified
Western Blot, Flow Cytometry, CyTOF
439728
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
EC 3.5.1.28, peptidoglycan recognition protein 2tagl-beta, peptidoglycan recognition protein L, PGLYRPLHMFT0141, PGRP-LN-acetylmuramoyl-L-alanine amidase, PGRPLPeptidoglycan recognition protein long, tagL, tagL-alpha, TAGL-like
Mouse myeloma cell line NS0-derived recombinant mouse PGLYRP2/PGRP-L, Ser23-Asn530, Accession # Q8VCS0
0.1 mL
Immunology, Inflammation
114770
Store at 4C in the dark.
IgG2a