missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD7 Antibody (46C713.2.1) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-56540JF635
This item is not returnable.
View return policy
Description
PDCD7 Monoclonal antibody specifically detects PDCD7 in Mouse samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| PDCD7 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| apoptosis-related protein ES18, ES18programmed cell death protein 7, FLJ54213, hES18, MGC22015, programmed cell death 7, U11/U12 snRNP 59K | |
| The antibody was raised against a synthetic peptide of mouse PDCD-7 protein. | |
| 0.1 mL | |
| Apoptosis | |
| 10081 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| 46C713.2.1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction