missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax3 Antibody (PAX3/4700) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20756JF585
This item is not returnable.
View return policy
Description
Pax3 Monoclonal antibody specifically detects Pax3 in Avian,Human samples. It is validated for ELISA
Specifications
| Pax3 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| CDHS, HuP2, HUP2MGC120384, MGC120381, MGC120382, MGC120383, MGC134778, paired box 3, paired box gene 3 (Waardenburg syndrome 1), paired box homeotic gene 3, paired box protein Pax-3, paired domain gene 3, paired domain gene HuP2, Waardenburg syndrome 1, WS1, WS3 | |
| Synthetic peptide (quail) from the C-terminus | |
| 0.1 mL | |
| Apoptosis, Mesenchymal Stem Cell Markers, Stem Cell Markers, Transcription Factors and Regulators | |
| 5077 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| ELISA | |
| PAX3/4700 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Avian, Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction