missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAR2 Antibody (Amgen patent anti-PAR-2) [Janelia Fluor™ 635], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28881JF635
This item is not returnable.
View return policy
Description
PAR2 Monoclonal antibody specifically detects PAR2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| PAR2 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| coagulation factor II (thrombin) receptor-like 1, Coagulation factor II receptor-like 1, GPR11protease-activated receptor 2, G-protein coupled receptor 11, PAR-2, PAR2G protein-coupled receptor-11, proteinase-activated receptor 2, proteinase-activated receptor-2, Thrombin receptor-like 1 | |
| PAR2 | |
| 0.1 mL | |
| GPCR, Signal Transduction | |
| 2150 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| Amgen patent anti-PAR-2 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction