missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pancreatic trypsin inhibitor Antibody (35), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-23590C
This item is not returnable.
View return policy
Description
Pancreatic trypsin inhibitor Monoclonal antibody specifically detects Pancreatic trypsin inhibitor in Bovine samples. It is validated for Western Blot, ELISA
Specifications
| Pancreatic trypsin inhibitor | |
| Monoclonal | |
| DyLight 650 | |
| Aprotinin, Basic protease inhibitor, BPI, BPTI, PTI, PTI pancreatic trypsin inhibitor | |
| Aprotinin from bovine lung | |
| 0.1 mL | |
| Primary | |
| Bovine | |
| Purified |
| Western Blot, ELISA | |
| 35 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 404172 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction