missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladin Antibody (1E6), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 16 publications
Brand: Novus Biologicals NBP1-25959
This item is not returnable.
View return policy
Description
Palladin Monoclonal specifically detects Palladin in Human, Mouse, Rat, Canine, Chicken samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications
| Palladin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 ug/ml, Flow Cytometry 5 ug/ml, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunoprecipitation 0.05 ml 2 ml lysate, Immunohistochemistry-Paraffin 1:50-1:100 | |
| CGI151, CGI-151, FLJ22190, FLJ38193, KIAA0992FLJ39139, MYN, myoneurin, palladin, palladin, cytoskeletal associated protein, PNCA1, Sarcoma antigen NY-SAR-77, SIH002FLJ61376 | |
| Mouse | |
| 85 kDa | |
| 0.1 mL | |
| Cellular Markers, Cytoskeleton Markers, Signal Transduction | |
| 23022 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1000000 | |
| Unconjugated | |
| Q8WX93 | |
| PALLD | |
| Endogenous Palladin immunoprecipitated from chicken embryo fibroblasts. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Chicken | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction