missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAAF1 Antibody (OTI1F11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73214CL1
This item is not returnable.
View return policy
Description
PAAF1 Monoclonal antibody specifically detects PAAF1 in Human,Mouse samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| PAAF1 | |
| Monoclonal | |
| CoraFluor 1 | |
| FLJ11848, PAAF, proteasomal ATPase-associated factor 1, Protein G-16, Rpn14, WD repeat domain 71, WD repeat-containing protein 71, WDR71 | |
| Full length human recombinant protein of human PAAF1(NP_079431) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI1F11 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 80227 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction