missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p23/PTGES3 Antibody (OTI2D2), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73186APC
This item is not returnable.
View return policy
Description
p23/PTGES3 Monoclonal antibody specifically detects p23/PTGES3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| p23/PTGES3 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI2D2 | |
| Western Blot, Immunohistochemistry | |
| cPGESp23, Cytosolic prostaglandin E2 synthase, Hsp90 co-chaperone, P23cytosolic prostaglandin E synthase, Progesterone receptor complex p23, prostaglandin E synthase 3, prostaglandin E synthase 3 (cytosolic), TEBPEC 5.3.99.3, Telomerase-binding protein p23, unactive progesterone receptor, 23 kD | |
| Full length human recombinant protein of human PTGES3 (NP_006592) produced in E.coli. | |
| 0.1 mL | |
| Membrane Trafficking and Chaperones | |
| 10728 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction