missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p18INK4c/CDKN2C Antibody (OTI2E1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73185CL1
This item is not returnable.
View return policy
Description
p18INK4c/CDKN2C Monoclonal antibody specifically detects p18INK4c/CDKN2C in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| p18INK4c/CDKN2C | |
| Monoclonal | |
| CoraFluor 1 | |
| CDK6 inhibitor p18, CDKN6, cyclin-dependent inhibitor, cyclin-dependent kinase 4 inhibitor C, Cyclin-dependent kinase 6 inhibitor, cyclin-dependent kinase inhibitor 2C (p18, inhibits CDK4), INK4Ccyclin-dependent kinase 6 inhibitor p18, p18, p18-INK4C, p18-INK6 | |
| Full length human recombinant protein of human CDKN2C (NP_001253) produced in E.coli. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 1031 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Western Blot | |
| OTI2E1 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction