missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXSM Antibody (OTI4E10), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73176PCP
This item is not returnable.
View return policy
Description
OXSM Monoclonal antibody specifically detects OXSM in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| OXSM | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI4E10 | |
| Western Blot, Immunohistochemistry | |
| 3-ketoacyl-acyl carrier protein synthase, 3-oxoacyl-[acyl-carrier-protein] synthase, mitochondrial, 3-oxoacyl-ACP synthase, mitochondrial, acyl-malonyl acyl carrier protein-condensing enzyme, beta-ketoacyl synthase, Beta-ketoacyl-ACP synthase, EC 2.3.1, EC 2.3.1.41, FASN2D, FLJ20604, KASI, KS3-oxoacyl-acyl-carrier-protein synthase, type II mitochondrial beta-ketoacyl synthase | |
| Human recombinant protein fragment corresponding to amino acids 78-343 of human OXSM (NP_060367) produced in E.coli. | |
| 0.1 mL | |
| metabolism | |
| 54995 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction