missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSMR beta Antibody (118125) [Janelia Fluor™ 669], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB662JF669
This item is not returnable.
View return policy
Description
OSMR beta Monoclonal antibody specifically detects OSMR beta in Mouse samples. It is validated for Western Blot,Immunocytochemistry
Specifications
| OSMR beta | |
| Monoclonal | |
| Janelia Fluor 669 | |
| IL-31 receptor subunit beta, IL-31R subunit beta, IL-31RB, IL-31R-beta, Interleukin-31 receptor subunit beta, MGC150627, MGC75127, oncostatin M receptor, oncostatin-M specific receptor beta subunit, oncostatin-M-specific receptor subunit beta, OSMR, OSMRB, OSMRBMGC150626, PLCA1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse OSM Rβ, Glu24-Leu738, Accession # O70458 | |
| RUO | |
| 9180 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Immunocytochemistry | |
| 118125 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction