missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCAPH Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-97603CL1
This item is not returnable.
View return policy
Description
NCAPH Polyclonal antibody specifically detects NCAPH in Human samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| NCAPH | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| barren (Drosophila) homolog, barren homolog (Drosophila), barren homolog 1, barren homolog 1 (Drosophila), Barren homolog protein 1, BRRN, BRRN1CAP-H, CAPH, Chromosome-associated protein H, condensin complex subunit 2, hCAP-H, KIAA0074, Non-SMC condensin I complex subunit H, non-SMC condensin I complex, subunit H, XCAP-H homolog | |
| Produced in rabbits immunized with E. coli-derived Human NCAPH fragment. | |
| 0.1 mL | |
| Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers | |
| 23397 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction