missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NALP6 Antibody (Clint-1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-11675CL1
This item is not returnable.
View return policy
Description
NALP6 Monoclonal antibody specifically detects NALP6 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Frozen)
Specifications
| NALP6 | |
| Monoclonal | |
| CoraFluor 1 | |
| Angiotensin II/Vasopressin Receptor, AVR, CLR11.4, NACHT, Leucine Rich Repeat And PYD Containing 6, NACHT, LRR And PYD Containing Protein 6, NACHT, LRR And PYD Domains-Containing Protein 6, NAVR, NAVR/AVR, NLR Family, Pyrin Domain Containing 6, NLRP6, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 6, PAN3, PYPAF5, PYRIN-Containing APAF1-Like Protein 5 | |
| Recombinant human NALP6 (NACHT domain). | |
| 0.1 mL | |
| Immunology, Inflammation | |
| 171389 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG κ |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen) | |
| Clint-1 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction