missing translation for 'onlineSavingsMsg'
Learn More

MZF1 Antibody (PCRP-MZF1-1E8) [Janelia Fluor™ 525], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP3-24013JF525

Product Code. 30171025

  • £491.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MZF1 Monoclonal antibody specifically detects MZF1 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MZF1
Monoclonal
Janelia Fluor 525
myeloid zinc finger 1, MZF1B, MZF-1zinc finger protein 42 (myeloid-specific retinoic acid-responsive), Zfp98, Zinc finger protein 42, ZNF42MZF, ZSCAN6Zinc finger and SCAN domain-containing protein 6
Recombinant fragment (around aa37-128) of human MZF1 protein
0.1 mL
Primary
Human
Purified
Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
PCRP-MZF1-1E8
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
7593
Store at 4°C in the dark.
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.