missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYLIP Antibody (OTI3A2), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72862F
This item is not returnable.
View return policy
Description
MYLIP Monoclonal antibody specifically detects MYLIP in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MYLIP | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI3A2 | |
| Western Blot | |
| band 4.1 superfamily member BZF1, BZF1, cellular modulator of immune recognition (c-MIR), EC 6.3.2.-, Idol, IDOLmyosin regulatory light chain-interacting protein, Inducible degrader of the LDL-receptor, MIRE3 ubiquitin-protein ligase MYLIP, myosin regulatory light chain interacting proteinE3 ubiquitin ligase-inducible degrader of the low density lipoprotein receptor | |
| Human recombinant protein fragment corresponding to amino acids 127-385 of human MYLIP(NP_037394) produced in E.coli. | |
| 0.1 mL | |
| Lipid and Metabolism, Zinc Finger | |
| 29116 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction