missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS34 Antibody (OTI4H2), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72788PCP
This item is not returnable.
View return policy
Description
MRPS34 Monoclonal antibody specifically detects MRPS34 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| MRPS34 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| OTI4H2 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| MGC2616, mitochondrial ribosomal protein S34, MRPS12, MRP-S12,28S ribosomal protein S34, mitochondrial, MRP-S34, S34mt | |
| Full length human recombinant protein of human MRPS34(NP_076425) produced in E.coli. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 65993 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction