missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL10 Antibody (OTI2B4), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72779APC
This item is not returnable.
View return policy
Description
MRPL10 Monoclonal antibody specifically detects MRPL10 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| MRPL10 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 124995 | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI2B4 | |
| Western Blot, Immunohistochemistry | |
| L10mt, MGC17973, mitochondrial ribosomal protein L10, MRP-L10L8mt, MRP-L839S ribosomal protein L10, mitochondrial, MRPL839S ribosomal protein L8, mitochondrial, RPML8L10MT | |
| Full length human recombinant protein of human MRPL10(NP_660298) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction