missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPP3 Antibody (OTI8F1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72771CL1
This item is not returnable.
View return policy
Description
MPP3 Monoclonal antibody specifically detects MPP3 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| MPP3 | |
| Monoclonal | |
| CoraFluor 1 | |
| Discs large homolog 3, discs, large homolog 3, DLG3MAGUK p55 subfamily member 3, membrane protein palmitoylated 3, membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3), Protein MPP3 | |
| Full length human recombinant protein of human MPP3 (NP_001923) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence | |
| OTI8F1 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 4356 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction