missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor III/Tissue Factor Antibody (CL3805) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61640AF532
This item is not returnable.
View return policy
Description
Coagulation Factor III/Tissue Factor Monoclonal antibody specifically detects Coagulation Factor III/Tissue Factor in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
Specifications
| Coagulation Factor III/Tissue Factor | |
| Monoclonal | |
| Alexa Fluor 532 | |
| CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), FLJ17960, TF, TFA, Thromboplastin, tissue factor | |
| This antibody was developed using a recombinant protein derived from P13726, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer | |
| 2152 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown | |
| CL3805 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction