missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIG2/Kindlin-2 Antibody (OTI9E4), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72687CL1
This item is not returnable.
View return policy
Description
MIG2/Kindlin-2 Monoclonal antibody specifically detects MIG2/Kindlin-2 in Canine,Human,Monkey,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| MIG2/Kindlin-2 | |
| Monoclonal | |
| CoraFluor 1 | |
| fermitin family homolog 2, fermitin family homolog 2 (Drosophila), fermitin family member 2, KIND2DKFZp686G11125, kindlin 2, Kindlin-2, mig-2, MIG2FLJ34213, mitogen inducible gene 2 protein, Mitogen-inducible gene 2 protein, PH domain-containing family C member 1, pleckstrin homology domain containing, family C (with FERM domain) member 1, pleckstrin homology domain containing, family C member 1, Pleckstrin homology domain-containing family C member 1, PLEKHC1FLJ44462, UNC112, UNC112B | |
| Full-length protein expressed in 293T cell transfected with human FERMT2 expression vector | |
| 0.1 mL | |
| Primary | |
| Canine, Human, Monkey, Mouse, Rat | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI9E4 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 10979 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction