missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGMT Antibody (OTI4F9), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72659APC
This item is not returnable.
View return policy
Description
MGMT Monoclonal antibody specifically detects MGMT in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| MGMT | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI4F9 | |
| Western Blot, Immunohistochemistry | |
| 6-O-methylguanine-DNA methyltransferase, EC 2.1.1.63, methylated-DNA--protein-cysteine methyltransferase, methylguanine-DNA methyltransferase, O-6-methylguanine-DNA methyltransferase, O6-methylguanine-DNA methyltransferase, O-6-methylguanine-DNA-alkyltransferase | |
| Full length human recombinant protein of human MGMT (NP_002403) produced in HEK293T cell. | |
| 0.1 mL | |
| Apoptosis, Cancer, Chromatin Research, Direct Reversal of DNA Damage, DNA Repair, Tumor Suppressors | |
| 4255 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction