missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mGluR7 Antibody (437310) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4405JF669
This item is not returnable.
View return policy
Description
mGluR7 Monoclonal antibody specifically detects mGluR7 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| mGluR7 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| FLJ40498, glutamate receptor, metabotropic 7, metabotropic glutamate receptor 7, mGlu7, mGluR7, MGLUR7GPRC1GGLUR7 | |
| HEK293 human embryonic kidney cell line transfected with human mGluR7, Gln35-Ile915, Accession # Q14831 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 437310 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Vision | |
| 2917 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction