missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEF2D Antibody (PCRP-MEF2D-3A4) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14120JF669
This item is not returnable.
View return policy
Description
MEF2D Monoclonal antibody specifically detects MEF2D in Human samples. It is validated for Flow Cytometry,Immunoprecipitation,Immunohistochemistry (Paraffin),Protein Array,Immunocytochemistry/ Immunofluorescence
Specifications
| MEF2D | |
| Monoclonal | |
| Janelia Fluor 669 | |
| DKFZp686I1536, MADS box transcription enhancer factor 2, polypeptide D (myocyte enhancerfactor 2D), MEF2D/DAZAP1 fusion, myocyte enhancer factor 2D, myocyte enhancer factor 2D/deleted in azoospermia associated protein 1 fusionprotein, myocyte-specific enhancer factor 2D | |
| Recombinant full-length human MEF2D protein (Uniprot: Q14814) | |
| 0.1 mL | |
| Phospho Specific | |
| 4209 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin), Protein Array, Immunocytochemistry/Immunofluorescence | |
| PCRP-MEF2D-3A4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction