missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-227AF594
This item is not returnable.
View return policy
Description
MAP1 Polyclonal antibody specifically detects MAP1 in Human samples. It is validated for Western Blot
Specifications
| MAP1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 594 | |
| MAP1, MAP-1PNMA4Paraneoplastic antigen Ma4, modulator of apoptosis 1, paraneoplastic antigen like 4 | |
| residues 337-351 of the human 39 kDa MAP-1 protein. | |
| 0.1 mL | |
| Apoptosis, MAP Kinase Signaling | |
| 64112 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction