Learn More
M-CSFR/CD115 Antibody (LY3022855) [Janelia Fluor™ 635], Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-28653JF635
Product Code. 30173070
  • £328.00 / 0.10mL
Description
M-CSFR/CD115 Monoclonal antibody specifically detects M-CSFR/CD115 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
M-CSFR/CD115
Monoclonal
Janelia Fluor 635
CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, Proto-oncogene c-Fms
CSF1R/ M-CSFR/ CD115
0.1 mL
Myeloid derived Suppressor Cell, Phospho Specific, Protein Kinase
1436
Store at 4°C in the dark.
IgG1
Flow Cytometry, ELISA, Functional Assay
LY3022855
50mM Sodium Borate
Human
Protein A purified
RUO
Primary
Human
Purified