missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRIG3 Antibody (814834) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3495JF585
This item is not returnable.
View return policy
Description
LRIG3 Monoclonal antibody specifically detects LRIG3 in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| LRIG3 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| FLJ90440, KIAA3016, leucine-rich repeats and immunoglobulin-like domains 3, leucine-rich repeats and immunoglobulin-like domains protein 3, LIG-3, LIG3FLJ26573, LIRG3 | |
| Mouse myeloma cell line NS0-derived recombinant human LRIG3, Asp28-Thr807, Accession # Q6UXM1 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Western Blot, CyTOF | |
| 814834 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 121227 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction