missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPAR5/LPA5/GPR92 Antibody (459812) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10297JF525
This item is not returnable.
View return policy
Description
LPAR5/LPA5/GPR92 Monoclonal antibody specifically detects LPAR5/LPA5/GPR92 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunocytochemistry,CyTOF
Specifications
| LPAR5/LPA5/GPR92 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| G protein-coupled receptor 92, GPR92G-protein coupled receptor 92, GPR93G-protein coupled receptor 93, KPG_010, LPA receptor 5, LPA5LPA-5, lysophosphatidic acid receptor 5 | |
| NS0 mouse myeloma cell line transfected with human LPAR5/LPA5/GPR92, Accession # Q9H1C0 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
| 459812 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR | |
| 57121 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction