missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPAR4/LPA4 Antibody (561203), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10217APCCY7
This item is not returnable.
View return policy
Description
LPAR4/LPA4 Monoclonal antibody specifically detects LPAR4/LPA4 in Human samples. It is validated for Flow Cytometry
Specifications
| LPAR4/LPA4 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2846 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry | |
| 561203 | |
| Flow Cytometry | |
| G protein-coupled receptor 23, GPR23P2Y purinoceptor 9, LPA4LPA-4, lysophosphatidic acid receptor 4, P2RY9G-protein coupled receptor 23, P2Y5-LIKE, P2Y5-like receptor, P2Y9LPA receptor 4, Purinergic receptor 9 | |
| Human embryonic kidney cell line HEK293-derived transfected with human LPAR4/LPA4, Accession # Q99677 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction