missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin alpha 3/Laminin-5 Antibody (546215) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB21441JF585
This item is not returnable.
View return policy
Description
Laminin alpha 3/Laminin-5 Monoclonal antibody specifically detects Laminin alpha 3/Laminin-5 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Flow Cytometry (Intracellular Staining),Immunocytochemistry
Specifications
| Laminin alpha 3/Laminin-5 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| a]3B;BM600;E170;LAMA3;Lama3B;LAMNA;LOCS | |
| Chinese hamster ovary cell line CHO-derived recombinant human Laminin alpha 3/Laminin-5, aa 21-1713, Accession # NP_000218 | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry | |
| 546215 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction