Learn More
Lactoferrin Antibody (002), mFluor Violet 500 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90704MFV500
This item is not returnable.
View return policy
Description
Lactoferrin Monoclonal antibody specifically detects Lactoferrin in Mouse samples. It is validated for ELISA
Specifications
| ELISA | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Lactoferrin (Accession#: AAH06904.1; Met1-Gln707). | |
| 0.1 mL | |
| IgG |
| 50mM Sodium Borate | |
| Protein A purified | |
| Primary |