Learn More
Kallikrein 15 Antibody (02), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26079AF700
Product Code. 30038093
  • £392.00 / 0.10mL
Description
Kallikrein 15 Monoclonal antibody specifically detects Kallikrein 15 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Kallikrein 15
Monoclonal
Alexa Fluor 700
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
02
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
ACO, ACO protease, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, HSRNASPH, kallikrein 15, kallikrein-15, kallikrein-like serine protease, kallikrein-related peptidase 15, prostinogen
Hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Kallikrein 15 (Accession#: NP_059979.2, Met1-Asn256)
0.1 mL
Cancer, Tumor Suppressors
55554
Store at 4C in the dark.
IgG1