missing translation for 'onlineSavingsMsg'
Learn More
Learn More
J Chain Antibody (Mc19-9), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64825C
This item is not returnable.
View return policy
Description
J Chain Monoclonal antibody specifically detects J Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Specifications
| J Chain | |
| Monoclonal | |
| DyLight 650 | |
| IgJ chain;Immunoglobulin J chain;immunoglobulin J polypeptide, linker protein for immunoglobulin alpha and mu polypeptides;J chain;Joining chain of multimeric IgA and IgM | |
| Human J Chain | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| Mc19-9 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction