missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 4/CD104 Antibody (450-9D), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65599MFV500
This item is not returnable.
View return policy
Description
Integrin beta 4/CD104 Monoclonal antibody specifically detects Integrin beta 4/CD104 in Human, Ferret, Mustelid samples. It is validated for Flow Cytometry
Specifications
| Integrin beta 4/CD104 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Ferret, Mustelid | |
| Purified |
| Flow Cytometry | |
| 450-9D | |
| Flow Cytometry | |
| CD104, CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, integrin, beta 4 | |
| Purified alpha6 beta4 integrin from A431 cells | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
| 3691 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction