missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-22R alpha 1 Antibody (305405) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2770JF635
This item is not returnable.
View return policy
Description
IL-22R alpha 1 Monoclonal antibody specifically detects IL-22R alpha 1 in Human samples. It is validated for Flow Cytometry,Immunocytochemistry,CyTOF
Specifications
| IL-22R alpha 1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| CRF2-9interleukin 22 receptor, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL22R, IL22R1, IL-22RA1, IL-22R-alpha-1, interleukin 22 receptor, alpha 1, interleukin-22 receptor subunit alpha-1, zcytoR11 | |
| Chinese hamster ovary cell line CHO-derived recombinant human IL-22 R alpha 1, Pro18-Thr228, Accession # Q8N6P7 | |
| 0.1 mL | |
| Cytokine Research, Immunology, Signal Transduction | |
| 58985 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 305405 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction