missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17E/IL-25 Antibody (Centocor patent anti-IL-25) [Janelia Fluor™ 635], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28161JF635
This item is not returnable.
View return policy
Description
IL-17E/IL-25 Monoclonal antibody specifically detects IL-17E/IL-25 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| IL-17E/IL-25 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| IL17E, IL-17E, IL-25, interleukin 25, Interleukin-17E, interleukin-25 | |
| IL-25 | |
| 0.1 mL | |
| Cytokine Research | |
| 64806 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Centocor patent anti-IL-25 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction