missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-12 p70/IL-12A Antibody (OTI2A8), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71031APC
This item is not returnable.
View return policy
Description
IL-12 p70/IL-12A Monoclonal antibody specifically detects IL-12 p70/IL-12A in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| IL-12 p70/IL-12A | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI2A8 | |
| Western Blot, Immunohistochemistry | |
| CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35 | |
| Human recombinant protein fragment corresponding to amino acids 57-253 of human IL12A(NP_000873) produced in E.coli. | |
| 0.1 mL | |
| Alzheimers Research, Immunology, Innate Immunity, Neurodegeneration | |
| 3592 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction