missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP-3 Antibody (IGFBP3/3423) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-24250JF669
This item is not returnable.
View return policy
Description
IGFBP-3 Monoclonal antibody specifically detects IGFBP-3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| IGFBP-3 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3 | |
| Recombinant human full-length IGFBP-3 protein | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins, Prostate Cancer | |
| 3486 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| IGFBP3/3423 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction