missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha 1 Antibody (2-48), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-08231CL1
This item is not returnable.
View return policy
Description
IFN-alpha 1 Monoclonal antibody specifically detects IFN-alpha 1 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Frozen),Immunocytochemistry/Immunofluorescence
Specifications
| IFN-alpha 1 | |
| Monoclonal | |
| CoraFluor 1 | |
| IFL;IFN;IFNA@;IFNA13;IFN-ALPHA;IFN-alphaD;Interferon alpha-1/13;leIF D | |
| Purified recombinant human IFN-alpha 1 (Uniprot: P01562) | |
| 0.1 mL | |
| Cancer, Immunology, Innate Immunity | |
| 3439 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence | |
| 1948-02-01 00:00:00 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction