missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYPE Antibody (OTI2A11), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71681F
This item is not returnable.
View return policy
Description
HYPE Monoclonal antibody specifically detects HYPE in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| HYPE | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI2A11 | |
| Western Blot | |
| AMPylator FICD, EC 2.7.7.n1, FIC domain containing, FIC domain-containing protein, fic S-phase protein cell division homolog, HIP-13, HIP13UNQ3041, huntingtin interacting protein 13, Huntingtin interacting protein E, huntingtin interactor protein E, Huntingtin yeast partner E, Huntingtin-interacting protein 13, Huntingtin-interacting protein E, HYPEadenosine monophosphate-protein transferase FICD, MGC5623 | |
| Human recombinant protein fragment corresponding to amino acids 61-265 of human FICD (NP_009007) produced in E.coli. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 11153 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction