missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF1 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32125APC
This item is not returnable.
View return policy
Description
HNF1 Polyclonal antibody specifically detects HNF1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
Specifications
| HNF1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| HNF1 homeobox A, IDDM20, LFB1transcription factor 1, hepatic, LF-B1, hepatic nuclear factor (HNF1), albuminproximal factor, MODY3, TCF1hepatic, Transcription factor 1 | |
| The immunogen this antibody was made to, maps to a region between residue 550 to 600 of human HNF1 Homeobox A using the numbering given in entry NP_000536.5 (GeneID 6927). | |
| 0.1 mL | |
| Transcription Factors and Regulators | |
| 6927 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| APC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction