missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3.3 Antibody (RM190), mFluor Violet 610 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26046MFV610
This item is not returnable.
View return policy
Description
Histone H3.3 Monoclonal antibody specifically detects Histone H3.3 in Vertebrate samples. It is validated for Western Blot, ELISA, Multiplex Immunoassay
Specifications
| Histone H3.3 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Vertebrates | |
| Purified |
| Western Blot, ELISA, Multiplex | |
| RM190 | |
| Western Blot, ELISA, Multiplex Immunoassay | |
| H3.3A, H3F3, H3F3A, Histone H3.3, Histone H3.3A, Histone H3.3B, MGC87782, MGC87783 | |
| A peptide corresponding to Human Histone H3.3 | |
| 0.1 mL | |
| Chromatin Research, Epigenetics | |
| 3020 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction