Learn More
GPRC5D Antibody (19), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26101UV
Product Code. 30027470
  • £427.00 / 0.10mL
Description
GPRC5D Monoclonal antibody specifically detects GPRC5D in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GPRC5D
Monoclonal
DyLight 350
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified
Flow Cytometry
19
Flow Cytometry
G protein-coupled receptor, family C, group 5, member D, G-protein coupled receptor family C group 5 member D, MGC129713, MGC129714, orphan G-protein coupled receptor
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human GPRC5D.
0.1 mL
GPCR
55507
Store at 4C in the dark.
IgG2b